Product Image
Thermo Scientific Chemicals

Carvedilol

CAS: 72956-09-3 | C24H26N2O4 | 406.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462450010
Price (EUR)
35,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
35,70
Each
Chemical Identifiers
CAS72956-09-3
IUPAC Name1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol
Molecular FormulaC24H26N2O4
InChI KeyOGHNVEJMJSYVRP-UHFFFAOYNA-N
SMILESCOC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point115°C to 119°C
Loss on drying=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only