Chemical Identifiers
CAS22662-39-1
IUPAC NameN-[3-chloro-4-(4-chlorophenoxy)phenyl]-2-hydroxy-3,5-diiodobenzamide
Molecular FormulaC19H11Cl2I2NO3
InChI KeyNEMNPWINWMHUMR-UHFFFAOYSA-N
SMILESOC1=C(I)C=C(I)C=C1C(=O)NC1=CC=C(OC2=CC=C(Cl)C=C2)C(Cl)=C1
View more