Product Image
Thermo Scientific Chemicals

Valnemulin hydrochloride

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4628300101 g
462832500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462830010
Price (EUR)
293,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
293,00
Each
Chemical Identifiers
CAS133868-46-9
IUPAC Name4-ethenyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-{[1-(2-amino-3-methylbutanamido)-2-methylpropan-2-yl]sulfanyl}acetate hydrochloride
Molecular FormulaC31H53ClN2O5S
InChI KeyMFBPRQKHDIVLOJ-UHFFFAOYNA-N
SMILESCl.CC(C)C(N)C(=O)NCC(C)(C)SCC(=O)OC1CC(C)(C=C)C(O)C(C)C23CCC(=O)C2C1(C)C(C)CC3
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotationon dry substance
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only