Product Image
Thermo Scientific Chemicals

Desloratadine

CAS: 100643-71-8 | C19H19ClN2 | 310.83 g/mol
Have Questions?
Catalog NumberQuantity
4629200101 g
Catalog number 462920010
Price (EUR)
167,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
167,00
Each
Chemical Identifiers
CAS100643-71-8
IUPAC Name13-chloro-2-(piperidin-4-ylidene)-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene
Molecular FormulaC19H19ClN2
InChI KeyJAUOIFJMECXRGI-UHFFFAOYSA-N
SMILESClC1=CC=C2C(CCC3=CC=CN=C3C2=C2CCNCC2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only