Zanamivir monohydrate
Thermo Scientific Chemicals

Zanamivir monohydrate

CAS: 1260601-68-0 | C12 H20 N4 O7 . H2 O | 350.33 g/mol
Have Questions?
Catalog NumberQuantity
4629600101 g
Catalog number 462960010
Price (EUR)
324,65
Online exclusive
360,00
Save 35,35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
324,65
Online exclusive
360,00
Save 35,35 (10%)
Each
Chemical Identifiers
CAS1260601-68-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Loss on drying4 % to 9 % (1 g, 105°C) (vacuum)
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Specific optical rotation(on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only