Product Image
Thermo Scientific Chemicals

Ritalinic acid

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 463000010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS19395-41-6
IUPAC Name2-phenyl-2-(piperidin-2-yl)acetic acid
Molecular FormulaC13H17NO2
InChI KeyINGSNVSERUZOAK-UHFFFAOYNA-N
SMILESOC(=O)C(C1CCCCN1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only