Strontium ranelate
Thermo Scientific Chemicals

Strontium ranelate

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 463011000
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS135459-87-9
IUPAC Namestrontium(2+) bis(2-{5-[bis(carboxymethyl)amino]-2-carboxy-4-cyanothiophen-3-yl}acetate)
Molecular FormulaC24H18N4O16S2Sr
InChI KeyKVJIIORUFXGTFN-UHFFFAOYSA-L
SMILES[Sr++].OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O.OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Water=<6 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only