Product Image
Thermo Scientific Chemicals

Tylosin tartrate

Have Questions?
Catalog NumberQuantity
4630700505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463070050
Price (EUR)
103,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
103,00
Each
Chemical Identifiers
CAS74610-55-2
IUPAC Name2,3-dihydroxybutanedioic acid; 2-[6-({5-[(4,5-dihydroxy-4,6-dimethyloxan-2-yl)oxy]-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl}oxy)-16-ethyl-4-hydroxy-15-{[(5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl)oxy]methyl}-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
Molecular FormulaC50H83NO23
InChI KeyICVKYYINQHWDLM-UHFFFAOYNA-N
SMILESOC(C(O)C(O)=O)C(O)=O.CCC1OC(=O)CC(O)C(C)C(OC2OC(C)C(OC3CC(C)(O)C(O)C(C)O3)C(C2O)N(C)C)C(CC=O)CC(C)C(=O)C=CC(C)=CC1COC1OC(C)C(O)C(OC)C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Solubility(5 % in water) Clear light yellow to yellow-green
HPLC>=94 % (sum of tylosins A+B+C+D)
Infrared spectrumConforms
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only