Product Image
Thermo Scientific Chemicals

Cefditoren pivoxyl

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4631100505 g
4631100101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463110050
Price (EUR)
320,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
320,00
Each
Chemical Identifiers
CAS117467-28-4
IUPAC Name{7-[2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[2-(4-methyl-1,3-thiazol-5-yl)ethenyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carbonyloxy}methyl 2,2-dimethylpropanoate
Molecular FormulaC25H28N6O7S3
InChI KeyAFZFFLVORLEPPO-UHFFFAOYNA-N
SMILESCON=C(C(=O)NC1C2SCC(C=CC3=C(C)N=CS3)=C(N2C1=O)C(=O)OCOC(=O)C(C)(C)C)C1=CSC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Water=<1.5 %
Appearance (Form)Powder
Titration with HClO494.0 to 106.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only