Product Image
Thermo Scientific Chemicals

Mosapride citrate

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463200010
Price (EUR)
98,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
98,30
Each
Chemical Identifiers
CAS112885-42-4
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid; 4-amino-5-chloro-2-ethoxy-N-({4-[(4-fluorophenyl)methyl]morpholin-2-yl}methyl)benzamide
Molecular FormulaC27H33ClFN3O10
InChI KeyHUZTYZBFZKRPFG-UHFFFAOYNA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O.CCOC1=CC(N)=C(Cl)C=C1C(=O)NCC1CN(CC2=CC=C(F)C=C2)CCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)powder
Infrared spectrumConforms
Water5 to 6 %
HPLC>=97.5 %
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only