Product Image
Thermo Scientific Chemicals

Pramipexole dihydrochloride monohydrate, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 463220050
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS191217-81-9
IUPAC Namedihydrogen (6S)-N6-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine hydrate dichloride
Molecular FormulaC10H21Cl2N3OS
InChI KeyAPVQOOKHDZVJEX-QTPLPEIMSA-N
SMILES[H+].[H+].O.[Cl-].[Cl-].CCCN[C@H]1CCC2=C(C1)SC(N)=N2
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Water4.5 % to 7.5 %
Optical rotation-63° to -69° (1 % in MeOH)
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only