Product Image
Thermo Scientific Chemicals

Repaglinide

CAS: 135062-02-1 | C27H36N2O4 | 452.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 463230010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS135062-02-1
IUPAC Name2-ethoxy-4-({[(1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]carbamoyl}methyl)benzoic acid
Molecular FormulaC27H36N2O4
InChI KeyFAEKWTJYAYMJKF-QHCPKHFHSA-N
SMILESCCOC1=CC(CC(=O)N[C@@H](CC(C)C)C2=CC=CC=C2N2CCCCC2)=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation+7° to +10° (20°C, 589 nm) (1% in MeOH)
Appearance (Color)White to almost white
Loss on drying=<0.5 % (105°C)
Appearance (Form)Powder
Melting point129°C to 135°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only