Drierite™, with indicator, 6 mesh
Thermo Scientific Chemicals

Drierite™, with indicator, 6 mesh

CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 463470020
Price (EUR)
256,00
Each
Quantity:
2 kg
Request bulk or custom format
Price (EUR)
256,00
Each
Chemical Identifiers
CAS7778-18-9
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue
Composition>=98 % (CaSO4) Typical
Water=<0.3 %
Composition=<2 % (CoCl2) Typical
Appearance (Form)Irregular granules
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only