Oxybutynin hydrochloride
Thermo Scientific Chemicals

Oxybutynin hydrochloride

CAS: 1508-65-2 | C22H32ClNO3 | 393.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 463500050
Price (EUR)
279,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
279,00
Each
Chemical Identifiers
CAS1508-65-2
IUPAC Name4-(diethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate hydrochloride
Molecular FormulaC22H32ClNO3
InChI KeySWIJYDAEGSIQPZ-UHFFFAOYNA-N
SMILESCl.CCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Loss on drying=<3 %
Titration with NaOH97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only