Product Image
Thermo Scientific Chemicals

Doripenem hydrate

Have Questions?
Catalog NumberQuantity
4638700101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463870010
Price (EUR)
212,65
Online exclusive
236,00
Save 23,35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
212,65
Online exclusive
236,00
Save 23,35 (10%)
Each
Chemical Identifiers
CAS1820954-21-9
IUPAC Name(4R,5S,6S)-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-3-{[(3S,5S)-5-[(sulfamoylamino)methyl]pyrrolidin-3-yl]sulfanyl}-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid
Molecular FormulaC15H24N4O6S2
InChI KeyAVAACINZEOAHHE-SEIFCSKWNA-N
SMILES[H][C@]12[C@@H](C)C(S[C@@H]3CN[C@H](CNS(N)(=O)=O)C3)=C(N1C(=O)[C@]2([H])[C@@H](C)O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Residue after ignition=<0.1 %
HPLC>=97.5 % (on dry substance)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only