Product Image
Thermo Scientific Chemicals

Ambrisentan, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 463950010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS177036-94-1
IUPAC Name2-[(4,6-dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropanoic acid
Molecular FormulaC22H22N2O4
InChI KeyOUJTZYPIHDYQMC-UHFFFAOYNA-N
SMILESCOC(C(OC1=NC(C)=CC(C)=N1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only