Product Image
Thermo Scientific Chemicals

Pseudopelletierine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 463980250
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS552-70-5
IUPAC Name9-methyl-9-azabicyclo[3.3.1]nonan-3-one
Molecular FormulaC9H15NO
InChI KeyRHWSKVCZXBAWLZ-UHFFFAOYNA-N
SMILESCN1C2CCCC1CC(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)White to yellow to brown
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only