Acetonitrile-d3, for NMR, 99.8 atom % D
Thermo Scientific Chemicals

Acetonitrile-d3, for NMR, 99.8 atom % D

CAS: 2206-26-0 | C2H3N | 44.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
25 mL
Catalog number 464160100
Price (EUR)
-
Quantity:
10 mL
Request bulk or custom format
Specifications
Boiling Point79.0°C to 81.0°C
CAS2206-26-0
Chemical Name or MaterialAcetonitrile-d3
Density0.8440 g/mL
EINECS Number218-616-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only