Product Image
Thermo Scientific Chemicals

Olanzapine

CAS: 132539-06-1 | C17H20N4S | 312.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464290010
Price (EUR)
73,65
Precio exclusivo en nuestra web
81,20
Save 7,55 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
73,65
Precio exclusivo en nuestra web
81,20
Save 7,55 (9%)
Each
Chemical Identifiers
CAS132539-06-1
IUPAC Name5-methyl-8-(4-methylpiperazin-1-yl)-4-thia-2,9-diazatricyclo[8.4.0.0³,⁷]tetradeca-1(14),2,5,7,10,12-hexaene
Molecular FormulaC17H20N4S
InChI KeyWXPNDRBBWZMPQG-UHFFFAOYSA-N
SMILESCN1CCN(CC1)C1=C2C=C(C)SC2=NC2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige
Appearance (Form)Powder or crystalline powder or crystals
Infrared spectrumConforms
Melting point192°C to 196°C
HPLC>=97.5 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only