4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals
4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals
4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-tert-Butyl-2,6-dimethylphenylsulfur trifluoride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 464350050
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS947725-04-4
IUPAC Name5-tert-butyl-1,3-dimethyl-2-(trifluoro-λ⁴-sulfanyl)benzene
Molecular FormulaC12H17F3S
InChI KeyVRTQPEYVMHATOA-UHFFFAOYSA-N
SMILESCC1=CC(=CC(C)=C1S(F)(F)F)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light red
Infrared spectrumConforms
Melting point62°C to 68°C
Appearance (Color)to light brown
Appearance (Form)Powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only