Product Image
Thermo Scientific Chemicals

Thiacloprid

CAS: 111988-49-9 | C10H9ClN4S | 252.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 464392500
Price (EUR)
60,80
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
60,80
Each
Chemical Identifiers
CAS111988-49-9
IUPAC Name({3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene}amino)formonitrile
Molecular FormulaC10H9ClN4S
InChI KeyHOKKPVIRMVDYPB-UHFFFAOYSA-N
SMILESClC1=CC=C(CN2CCSC2=NC#N)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
HPLC>=95.0 %
Water=<1 %
Appearance (Color)Powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only