Product Image
Thermo Scientific Chemicals

Adapalene

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 464500050
Price (EUR)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS106685-40-9
IUPAC Name6-[3-(adamantan-1-yl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
Molecular FormulaC28H28O3
InChI KeyLZCDAPDGXCYOEH-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1C12CC3CC(CC(C3)C1)C2)C1=CC=C2C=C(C=CC2=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Loss on drying=<1 % (105°C, 4 h)
Appearance (Form)Crystalline powder
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only