Silodosin, Thermo Scientific Chemicals
Product Image
Thermo Scientific Chemicals

Silodosin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464521000
Price (EUR)
62,10
Each
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
62,10
Each
Add to cart
Chemical Identifiers
CAS160970-54-7
IUPAC Name1-(3-hydroxypropyl)-5-[(2R)-2-({2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl}amino)propyl]-2,3-dihydro-1H-indole-7-carboxamide
Molecular FormulaC25H32F3N3O4
InChI KeyPNCPYILNMDWPEY-QGZVFWFLSA-N
SMILESC[C@H](CC1=CC2=C(N(CCCO)CC2)C(=C1)C(N)=O)NCCOC1=CC=CC=C1OCC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<0.3 %
Residue after ignition=<0.1 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only