Product Image
Thermo Scientific Chemicals

Tetramethylrhodamine methyl ester perchlorate

CAS: 115532-50-8 | C25H25ClN2O7 | 500.93 g/mol
Have Questions?
Catalog NumberQuantity
46456025025 mg
Catalog number 464560250
Price (EUR)
177,65
線上優惠
197,00
Save 19,35 (10%)
Each
Quantity:
25 mg
Request bulk or custom format
Price (EUR)
177,65
線上優惠
197,00
Save 19,35 (10%)
Each
Chemical Identifiers
CAS115532-50-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
Appearance (Form)Crystals
Infrared spectrumConforms
HPLC>=94 %
Molar absorption coefficient>=95000 l/molcm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only