Product Image
Thermo Scientific Chemicals

Dapagliflozin

CAS: 461432-26-8 | C21H25ClO6 | 408.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 464620010
Price (EUR)
110,65
キャンペーン価格
123,00
Save 12,35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
110,65
キャンペーン価格
123,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS461432-26-8
IUPAC Name2-{4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC21H25ClO6
InChI KeyJVHXJTBJCFBINQ-UHFFFAOYNA-N
SMILESCCOC1=CC=C(CC2=C(Cl)C=CC(=C2)C2OC(CO)C(O)C(O)C2O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Water=<2 %
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only