Product Image
Thermo Scientific Chemicals

Ginsenoside Rb1

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Catalog number 464630500
Price (EUR)
110,00
Each
Quantity:
50 mg
Request bulk or custom format
Price (EUR)
110,00
Each
Chemical Identifiers
CAS41753-43-9
IUPAC Name(2S,3R,4S,5S,6R)-2-({2-[(1S,3aR,3bR,5aR,7S,9aR,9bR,11R,11aR)-7-{[(2R,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}-11-hydroxy-3a,3b,6,6,9a-pentamethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-6-methylhept-5-en-2-yl}oxy)-6-({[(3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)oxane-3,4,5-triol
Molecular FormulaC54H92O23
InChI KeyGZYPWOGIYAIIPV-NGBMAODDSA-N
SMILESCC(C)=CCCC(C)(O[C@@H]1O[C@H](COC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@H](O)[C@H]1O)[C@H]1CC[C@]2(C)[C@@H]1[C@H](O)C[C@@H]1[C@@]3(C)CC[C@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)C(C)(C)[C@@H]3CC[C@@]21C
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Water=<10 %
Solubilitylight yellow
Appearance (Color)White
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only