Product Image
Thermo Scientific Chemicals

Succinimidyl acetate

CAS: 14464-29-0 | C6H7NO4 | 157.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 464750250
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS14464-29-0
IUPAC Name2,5-dioxopyrrolidin-1-yl acetate
Molecular FormulaC6H7NO4
InChI KeySIFCHNIAAPMMKG-UHFFFAOYSA-N
SMILESCC(=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<2 %
Appearance (Color)White to almost white
Melting point132°C to 136°C
HPLC>=97.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only