Product Image
Thermo Scientific Chemicals

Rocuronium bromide, 97%

CAS: 119302-91-9 | C32H53BrN2O4 | 609.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465022500
Price (EUR)
95,30
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
95,30
Each
Chemical Identifiers
CAS119302-91-9
IUPAC Name1-[(1R,2S,3aS,3bR,5aS,7S,8S,9aS,9bS,11aS)-1-(acetyloxy)-7-hydroxy-9a,11a-dimethyl-8-(morpholin-4-yl)-hexadecahydro-1H-cyclopenta[a]phenanthren-2-yl]-1-(prop-2-en-1-yl)pyrrolidin-1-ium bromide
Molecular FormulaC32H53BrN2O4
InChI KeyOYTJKRAYGYRUJK-FMCCZJBLSA-M
SMILES[Br-].CC(=O)O[C@H]1[C@H](C[C@H]2[C@@H]3CC[C@H]4C[C@H](O)[C@H](C[C@]4(C)[C@H]3CC[C@]12C)N1CCOCC1)[N+]1(CC=C)CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<6 %
Appearance (Color)White to light yellow to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO496.0 to 104.0 % (dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only