Product Image
Thermo Scientific Chemicals

Balsalazide disodium dihydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4650900505 g
4650900101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465090050
Price (EUR)
152,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
152,00
Each
Chemical Identifiers
CAS150399-21-6
IUPAC Namedisodium (3Z)-3-(2-{4-[(2-carboxylatoethyl)carbamoyl]phenyl}hydrazin-1-ylidene)-6-oxocyclohexa-1,4-diene-1-carboxylate dihydrate
Molecular FormulaC17H17N3Na2O8
InChI KeyHYSVUPBQPCRAJM-PIFVRXTCSA-L
SMILESO.O.[Na+].[Na+].[O-]C(=O)CCNC(=O)C1=CC=C(N\N=C2\C=CC(=O)C(=C2)C([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-red
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Water7 to 10 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only