Product Image
Thermo Scientific Chemicals

Schisandrin B, 96%, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
465142500250 mg
Catalog number 465142500
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS61281-37-6
IUPAC Name3,4,5,19-tetramethoxy-9,10-dimethyl-15,17-dioxatetracyclo[10.7.0.0²,⁷.0¹⁴,¹⁸]nonadeca-1(19),2(7),3,5,12,14(18)-hexaene
Molecular FormulaC23H28O6
InChI KeyRTZKSTLPRTWFEV-UHFFFAOYNA-N
SMILESCOC1=CC2=C(C(OC)=C1OC)C1=C(OC)C3=C(OCO3)C=C1CC(C)C(C)C2
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=95.0 %
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<3 %
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only