Product Image
Thermo Scientific Chemicals

1-Isoindolinone

CAS: 480-91-1 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 465180010
Price (EUR)
52,80
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
52,80
Each
Chemical Identifiers
CAS480-91-1
IUPAC Name2,3-dihydro-1H-isoindol-1-one
Molecular FormulaC8H7NO
InChI KeyPXZQEOJJUGGUIB-UHFFFAOYSA-N
SMILESO=C1NCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point149°C to 153°C
Infrared spectrumConforms
Appearance (Color)White to light yellow
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only