Product Image
Thermo Scientific Chemicals

Acacetin

CAS: 480-44-4 | C16H12O5 | 284.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465232500
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS480-44-4
IUPAC Name5,7-dihydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one
Molecular FormulaC16H12O5
InChI KeyDANYIYRPLHHOCZ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<3 %
Appearance (Color)White to light yellow to light brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only