Product Image
Thermo Scientific Chemicals

Irbesartan

CAS: 138402-11-6 | C25H28N6O | 428.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 465250050
Price (EUR)
172,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
172,00
Each
Chemical Identifiers
CAS138402-11-6
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<2 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point180°C to 184°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only