Fluocinonide
Thermo Scientific Chemicals

Fluocinonide

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
465452500250 mg
4654500101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465452500
Price (EUR)
195,65
キャンペーン価格
217,00
Save 21,35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
195,65
キャンペーン価格
217,00
Save 21,35 (10%)
Each
Chemical Identifiers
CAS356-12-7
IUPAC Name2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosa-14,17-dien-8-yl]-2-oxoethyl acetate
Molecular FormulaC26H32F2O7
InChI KeyWJOHZNCJWYWUJD-IUGZLZTKSA-N
SMILESCC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]1[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]21C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<1 %
HPLC>=96.0 %
Appearance (Color)White to beige to light brown
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only