Lupeol
Lupeol
Lupeol
Thermo Scientific Chemicals

Lupeol

CAS: 545-47-1 | C30H50O | 426.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465502500
Price (EUR)
269,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
269,00
Each
Chemical Identifiers
CAS545-47-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=93 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only