Product Image
Thermo Scientific Chemicals

Maleic anhydride, briquettes

CAS: 108-31-6 | C4H2O3 | 98.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
25 kg
Catalog number 465660050
Price (EUR)
87,10
Each
Quantity:
5 kg
Request bulk or custom format
Price (EUR)
87,10
Each
Specifications
CAS108-31-6
Chemical Name or MaterialMaleic anhydride
EINECS Number203-571-6
Flash Point102°C
Formula Weight98.06
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only