Product Image
Thermo Scientific Chemicals

Crizotinib

CAS: 877399-52-5 | C21H22Cl2FN5O | 450.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465692500
Price (EUR)
135,65
線上優惠
150,00
Save 14,35 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
135,65
線上優惠
150,00
Save 14,35 (10%)
Each
Chemical Identifiers
CAS877399-52-5
IUPAC Name3-[1-(2,6-dichloro-3-fluorophenyl)ethoxy]-5-[1-(piperidin-4-yl)-1H-pyrazol-4-yl]pyridin-2-amine
Molecular FormulaC21H22Cl2FN5O
InChI KeyKTEIFNKAUNYNJU-UHFFFAOYNA-N
SMILESCC(OC1=C(N)N=CC(=C1)C1=CN(N=C1)C1CCNCC1)C1=C(Cl)C=CC(F)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to beige
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only