Etodolac
Thermo Scientific Chemicals

Etodolac

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465720250
Price (EUR)
126,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
126,00
Each
Chemical Identifiers
CAS41340-25-4
IUPAC Name2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
Molecular FormulaC17H21NO3
InChI KeyNNYBQONXHNTVIJ-UHFFFAOYNA-N
SMILESCCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 %
HPLC>=97.5 %
Solubility(10 mg/ml in methanol) Clear colorless to yellow
Appearance (Color)White to light yellow to light beige
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only