Betamethasone sodium phosphate
Betamethasone sodium phosphate
Thermo Scientific Chemicals

Betamethasone sodium phosphate

Have Questions?
Catalog NumberQuantity
4657400101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465740010
Price (EUR)
101,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
101,00
Each
Add to cart
Chemical Identifiers
CAS151-73-5
IUPAC Namedisodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular FormulaC22H28FNa2O8P
InChI KeyPLCQGRYPOISRTQ-LWCNAHDDSA-L
SMILES[Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Specific optical rotation+98° to +105° (20°C, 589 nm) (1% in water)
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only