Fulvestrant
Fulvestrant
Fulvestrant
Thermo Scientific Chemicals

Fulvestrant

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
46578050050 mg
4657800101 g
465782500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465780500
Price (EUR)
126,65
Online Exclusive
140,00
Save 13,35 (10%)
Each
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (EUR)
126,65
Online Exclusive
140,00
Save 13,35 (10%)
Each
Add to cart
Chemical Identifiers
CAS129453-61-8
IUPAC Name(1S,3aS,3bR,4R,9bS,11aS)-11a-methyl-4-[9-(4,4,5,5,5-pentafluoropentanesulfinyl)nonyl]-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol
Molecular FormulaC32H47F5O3S
InChI KeyVWUXBMIQPBEWFH-WCCTWKNTSA-N
SMILESC[C@]12CC[C@H]3[C@H]([C@@H]1CC[C@@H]2O)[C@H](CCCCCCCCCS(=O)CCCC(F)(F)C(F)(F)F)CC1=CC(O)=CC=C31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=98.5 %
Solubility(0.1g in 10 ml Ethanol) Clear solution
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only