2,2'-Biimidazole
2,2'-Biimidazole
2,2'-Biimidazole
Thermo Scientific Chemicals

2,2'-Biimidazole

CAS: 492-98-8 | C6H6N4 | 134.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 465820010
Price (EUR)
98,40
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
98,40
Each
Chemical Identifiers
CAS492-98-8
IUPAC Name1H,1'H-2,2'-biimidazole
Molecular FormulaC6H6N4
InChI KeyAZUHIVLOSAPWDM-UHFFFAOYSA-N
SMILESN1C=CN=C1C1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to brown to grey
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Titration with HClO497.5 to 102.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only