Bedaquiline fumarate
Thermo Scientific Chemicals

Bedaquiline fumarate

CAS: 845533-86-0 | C36H35BrN2O6, C32H31BrN2O2·C4H4O4 | 671.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465912500
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS845533-86-0
IUPAC Name1-(6-bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol; but-2-enedioic acid
Molecular FormulaC36H35BrN2O6
InChI KeyZLVSPMRFRHMMOY-UHFFFAOYNA-N
SMILESOC(=O)C=CC(O)=O.COC1=C(C=C2C=C(Br)C=CC2=N1)C(C1=CC=CC=C1)C(O)(CCN(C)C)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<1 %
Appearance (Color)White to almost white
Appearance (Form)Powder
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only