Diethylthiocarbamoyl chloride, Thermo Scientific Chemicals
Diethylthiocarbamoyl chloride, Thermo Scientific Chemicals
Diethylthiocarbamoyl chloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethylthiocarbamoyl chloride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 465920050
Price (EUR)
58,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
58,30
Each
Chemical Identifiers
CAS88-11-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Solid
Infrared spectrumConforms
Melting point45°C to 50°C
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only