Rutaecarpine
Thermo Scientific Chemicals

Rutaecarpine

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465980010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS84-26-4
IUPAC Name3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-1(21),2(10),4,6,8,15,17,19-octaen-14-one
Molecular FormulaC18H13N3O
InChI KeyACVGWSKVRYFWRP-UHFFFAOYSA-N
SMILESO=C1N2CCC3=C(NC4=CC=CC=C34)C2=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to beige
Appearance (Form)Powder
Loss on drying=<0.5 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only