Dimethyl (1-diazo-2-oxopropyl)phosphonate, 95%
Dimethyl (1-diazo-2-oxopropyl)phosphonate, 95%
Thermo Scientific Chemicals

Dimethyl (1-diazo-2-oxopropyl)phosphonate, 95%

CAS: 90965-06-3 | C5H9N2O4P | 192.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 466020010
Price (EUR)
103,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
103,00
Each
Add to cart
Chemical Identifiers
CAS90965-06-3
IUPAC Name1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate
Molecular FormulaC5H9N2O4P
InChI KeySQHSJJGGWYIFCD-UHFFFAOYSA-N
SMILESCOP(=O)(OC)C([N+]#N)=C(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94 %
Appearance (Color)Clear light yellow to brown
Appearance (Form)liquid
Infrared spectrumConforms
Refractive index1.4770 to 1.4820 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only