Nelfinavir mesylate
Thermo Scientific Chemicals

Nelfinavir mesylate

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
466202500250 mg
4662000101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466202500
Price (EUR)
651,00
Each
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
651,00
Each
Chemical Identifiers
CAS159989-65-8
IUPAC Name(3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylphenyl)formamido]-4-(phenylsulfanyl)butyl]-decahydroisoquinoline-3-carboxamide; methanesulfonic acid
Molecular FormulaC33H49N3O7S2
InChI KeyNQHXCOAXSHGTIA-SKXNDZRYSA-N
SMILESCS(O)(=O)=O.CC1=C(O)C=CC=C1C(=O)N[C@@H](CSC1=CC=CC=C1)[C@H](O)CN1C[C@H]2CCCC[C@H]2C[C@H]1C(=O)NC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Specific optical rotation-101° to -115° (20°C, 589 nm) (1% in Methanol) (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only