Olaparib
Thermo Scientific Chemicals

Olaparib

CAS: 763113-22-0 | C24H23FN4O3, C24H23FN4O3 | 434.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 466290010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS763113-22-0
IUPAC Name4-{[3-(4-cyclopropanecarbonylpiperazine-1-carbonyl)-4-fluorophenyl]methyl}-1,2-dihydrophthalazin-1-one
Molecular FormulaC24H23FN4O3
InChI KeyFDLYAMZZIXQODN-UHFFFAOYSA-N
SMILESFC1=C(C=C(CC2=NNC(=O)C3=CC=CC=C23)C=C1)C(=O)N1CCN(CC1)C(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)crystalline powder
Infrared spectrumcomplies
HPLC>=98.0 %
Water=<1.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only