Raloxifene hydrochloride
Thermo Scientific Chemicals

Raloxifene hydrochloride

CAS: 82640-04-8 | C28H28ClNO4S, C28H27NO4S·HCl | 510.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466500050
Price (EUR)
567,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
567,00
Each
Chemical Identifiers
CAS82640-04-8
IUPAC Namehydrogen 2-(4-hydroxyphenyl)-3-{4-[2-(piperidin-1-yl)ethoxy]benzoyl}-1-benzothiophen-6-ol chloride
Molecular FormulaC28H28ClNO4S
InChI KeyBKXVVCILCIUCLG-UHFFFAOYSA-N
SMILES[H+].[Cl-].OC1=CC=C(C=C1)C1=C(C(=O)C2=CC=C(OCCN3CCCCC3)C=C2)C2=CC=C(O)C=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point265°C to 270°C
Loss on drying=<0.5 % (105°C, 3 hr)
HPLC>=97.5 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only