Efavirenz
Thermo Scientific Chemicals

Efavirenz

CAS: 154598-52-4 | C14H9ClF3NO2, C14H9ClF3NO2 | 315.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466620250
Price (EUR)
180,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
180,00
Each
Add to cart
Chemical Identifiers
CAS154598-52-4
IUPAC Name(4S)-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-2,4-dihydro-1H-3,1-benzoxazin-2-one
Molecular FormulaC14H9ClF3NO2
InChI KeyXPOQHMRABVBWPR-ZDUSSCGKSA-N
SMILESFC(F)(F)[C@]1(OC(=O)NC2=CC=C(Cl)C=C12)C#CC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)crystalline powder
HPLC>=98.0 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Water=<2.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only