Flurprimidol
Thermo Scientific Chemicals

Flurprimidol

CAS: 56425-91-3 | C15H15F3N2O2, C15H15F3N2O2 | 312.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466635000
Price (EUR)
261,00
Each
Quantity:
500 mg
Request bulk or custom format
Price (EUR)
261,00
Each
Chemical Identifiers
CAS56425-91-3
IUPAC Name2-methyl-1-(pyrimidin-5-yl)-1-[4-(trifluoromethoxy)phenyl]propan-1-ol
Molecular FormulaC15H15F3N2O2
InChI KeyVEVZCONIUDBCDC-UHFFFAOYNA-N
SMILESCC(C)C(O)(C1=CC=C(OC(F)(F)F)C=C1)C1=CN=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point92.0°C to 99.0°C
Water=<1.0 %
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
Appearance (Form)crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only